Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Papanicolaou Stain, EA-36-50 Formula, Ricca Chemical
SDP

Catalog No. 550016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5500-16 500 mL Natural Poly Bottle
5500-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5500-16 Supplier Ricca Chemical Company Supplier No. 550016

Counterstain for use with Hematoxylin Stains in Papanicolaou Staining

Specifications

Chemical Name or Material Papanicolaou Stain
Name Note EA-36-50 Formula
CAS 64-17-5, 7732-18-5, 67-56-1, 17372-87-1, 12501-23-4, 10114-58-6
CAS Min % 90.0, 6.0, 0.2, 0.23, 0.06, 9.0E-4
CAS Max % 91.0, 8.0, 0.3, 0.24, 0.06, 9.0E-4
Grade Laboratory
Packaging Natural Poly Bottle
Color Green
pH 6 to 7
Quantity 500 mL
Physical Form Murky Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.