Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAPD5/TRF4-2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt.
PAPD5 plays a role in replication-dependent histone mRNA degradation.It may be involved in the terminal uridylation of mature histone mRNAs before their degradation is initiated. DNA polymerase, probably involved in DNA repair. It may play a role in sister chromatid cohesion.
Spécifications
Spécifications
| Antigen | PAPD5/TRF4-2 |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Formulation | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gene | PAPD5 |
| Gene Accession No. | Q8NDF8 |
| Gene Alias | PAP associated domain containing 5 |
| Gene Symbols | PAPD5 |
| Afficher plus de résultats |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'