Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Paper Board Sign: CAUTION - HOT

Catalog No. 19104870 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
10 x 7 in.
14 x 10 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-104-870 14 x 10 in.
19-104-871 10 x 7 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-104-870 Supplier Brady™ Supplier No. 116068

Improve workplace safety with proper signage and markings. This Brady™ paper board sign is a durable, clear and effective tool to raise awareness to workplace hazards.

  • Paper board material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use

Specifications

Material Paper Board
Shape Rectangular
Dimensions (L x W) 14 x 10 in.
Displayed Text CAUTION - HOT
Color Black on Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.