Learn More
Essendant PAPER ROLLMULTI64X300
Supplier:  Essendant EPSS045452N
Encompass
The Epson DS Transfer Multi Purpose paper is an extremely high quality excellent wide color gamut water-based sublimation transfer paper It s finished output provides high ink transferability on an extremely low cockling base The paper is optimized for transferring onto multiple sublimation substrates This transfer paper is designed to deliver the best quality when used with Epson ink Sheet/Roll Size- 64x300 ft Paper Grade (gsm)- 105 gsm Paper/Media Thickness (mil)- 5 3 mil Paper/Media Color(s)- White
Catalog No. 50-000-35578
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.