Learn More
Essendant PAPER24X100MTLCLSTRWH
Supplier:  Essendant EPSS045593
Encompass
The Epson Professional Media Metallic Luster Photo Paper is designed with an incredible Dmax and rich color gamut With its resin coated paper base this semi-gloss E surface photographic paper features a unique metallic surface that provides extreme sharpness and depth It is designed to deliver the best in photographic image quality and longevity when used with Epson ink Sheet/Roll Size- 24x100 ft Paper/Media Thickness (mil)- 10 5 mil Paper/Media Color(s)- White Color Family- White
Catalog No. 50-000-34991
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.