Learn More
Essendant PAPERENHNCDSPR-BMT
Supplier:  Essendant EPSS041339
Encompass_Preferred
For photographers seeking a flat matte surface Ultra Premium Presentation Paper Matte is the ideal choice This bright white paper is perfect for images that do not require gloss It yields highly saturated images while maintaining excellent highlight and shadow detail Designed primarily for use with UltraChrome Inks EPSON Archival Inks and DuraBrite Inks this versatile paper can also be used with dye-based inks Hobbyists professional photographers and digital artists alike will find Epson Ultra Premium Presentation Paper Matte a remarkable solution for any project Sheet/Roll Size- 13x19 Paper/Media Thickness (mil)- 10 mil Paper/Media Color(s)- White Color Family- White
Catalog No. 50-000-32461