Learn More
Essendant PAPERHOT PRESS BRIGHT
Supplier:  Essendant EPSS042331
Encompass_Preferred
For fine art photographers looking for a matte medium that can deliver a wide color gamut and high brightness Epson has developed Hot Press Bright Paper Made from 100pct cotton rag and using a hot press production technique this paper features a smooth matte finish The heavy weight of 330 gsm thickness of 18 mil and opacity of 98pct further give this paper the look and feel of a true fine art paper Also through the use of optical brightening agents this media will help deliver bright whites deep blacks and a wide color gamut Designed for longevity Hot Press Bright is free of acid lignin and chlorine and is pH buffered Sheet/Roll Size- 17x22 Paper/Media Thickness (mil)- 17 mil Paper/Media Color(s)- Bright White Color Family- White
Catalog No. 50-000-35196
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.