Learn More
Essendant PAPERLCGY PLTN17X50WH
Supplier:  Essendant EPSS450076
Encompass
Legacy Platine is a 100pct cotton fibre paper with a white smooth satin finish This paper combines the look and feel of the revered silver halide F-surface with a true cotton base It has an industry leading microporous inkjet receptive layer that produces deep rich blacks (Dmax) expanded color gamut and gentle tonal gradations and will provide outstanding image permanence and durability Sheet/Roll Size- 17x50 ft Paper/Media Thickness (mil)- 17 mil Paper/Media Color(s)- White Color Family- White
Catalog No. 50-000-34949
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.