Learn More
Essendant PAPERPOSTRBRD24X3610PK
Supplier:  Essendant EPSS450433
Encompass
For photographers seeking a flat matte surface with the stiffness of a mounted product Epson Enhanced Matte Posterboard is the ideal choice This bright white posterboard is perfect for short term signage and display It yields highly saturated images while maintaining excellent highlight and shadow detail Designed primarily for use with Epson UltraChrome and Archival Inks this versatile posterboard can also be used with dye-based inks Hobbyists professional photographers and digital artists alike will find Epson Enhanced Matte Posterboard a remarkable solution for any project Art Board Type- Flat Railroad and Poster Board Material(s)- Alpha-cellulose Paper Color(s)- White Board Size Open (WxH)- 24x36
Catalog No. 50-000-35248