Learn More
PAPOLA Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301009AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
Polymerase that creates the 3′-poly(A) tail of mRNA′s. Also required for the endoribonucleolytic cleavage reaction at some polyadenylation sites. May acquire specificity through interaction with a cleavage and polyadenylation specificity factor (CPSF) at its C-terminus.Specifications
| PAPOLA | |
| Polyclonal | |
| Unconjugated | |
| PAPOLA | |
| PAP, PAP-alpha, polynucleotide adenylyltransferase alpha | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10914 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| P51003 | |
| PAPOLA | |
| Between 600 and 650 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.