Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAPPA Monoclonal Antibody (PAPP A 1-24)
GREENER_CHOICE

Catalog No. ABS0062402
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS0062402 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS0062402 Supplier Thermo Scientific Supplier No. ABS0062402
Only null left
Add to Cart
Add to Cart

PAPPA Monoclonal Antibody (PAPP A 1-24)

TRUSTED_SUSTAINABILITY

Specifications

Antigen PAPPA
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone PAPP A 1-24
Concentration 1.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene PAPPA
Gene Accession No. Q13219
Gene Alias 8430414N03Rik; ASBABP2; aspecific BCL2 ARE-binding protein 2; differentially placenta 1 expressed protein; DIPLA1; IGFBP-4 ase; IGFBP-4ase; IGF-dependent IGFBP-4 protease; insulin-like growth factor-dependent IGF binding protein-4 protease; insulin-like growth factor-dependent IGF-binding protein 4 protease; PAG1; PAPA; PAPP A; PAPPA; PAPP-A; PAPPA1; Pappalysin; pappalysin 1; pappalysin-1; pregnacy-associated plasma protein A; pregnancy-associated plasma protein A; pregnancy-associated plasma protein A, pappalysin 1; RP11-45A16.1
Gene Symbols PAPPA
Host Species Mouse
Immunogen Purified human PAPP-A.
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5069
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.