Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA PAPR Double Bib Hood

Catalog No. 19177178 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
OptimAir™ TL PAPR
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-177-178 OptimAir™ TL PAPR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-177-178 Supplier MSA Supplier No. 10083330
Add to Cart
Add to Cart

For Use With OptimAir™ TL PAPR

Includes threaded connector with 1-touch adjustable suspension.

Specifications

Certifications/Compliance NIOSH
Color White
Material Tychem QC
Size One Size
For Use With (Equipment) OptimAir™ TL PAPR
Model 10083330
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.