Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ PAPR Headgear Sweatband

Catalog No. 19085571 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Universal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-085-571 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-085-571 Supplier Honeywell™ Supplier No. PA104
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with Primair™ PAPR systems.

  • Sweatband
  • For use with Primair Series
  • Comfort and absorbent
  • Pack of 10

Specifications

Color Black
Disposable Reusable
Product Type Sweatband
Size One Size
For Use With (Equipment) Primair™ PAPR Systems
Model PA104
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.