Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAQR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566990
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566990 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566990 Supplier Invitrogen™ Supplier No. PA566990
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YHLFMCHQGGS AVYARLLALD M Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.

PAQR4 gene ontology annotations for this gene involves signaling receptor activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAQR4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PAQR4
Gene Accession No. Q8N4S7
Gene Alias 1500004C10Rik; LOW QUALITY PROTEIN: progestin and adipoQ receptor family member 4; MNCb-0914; Paqr4; Progestin and adipoQ receptor family member 4; progestin and adipoQ receptor family member IV
Gene Symbols PAQR4
Host Species Rabbit
Immunogen Recombinant Human PAQR4. Recombinant protein control fragment (Product #RP-107633).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124222
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.