Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 85315 |
| Product Line | Novus |
| Purification Method | >80% by SDS-PAGE and Coomassie blue staining |
| Common Name | PAQR8 Recombinant Protein Antigen |
| Content And Storage | Store at −20°C. Avoid freeze-thaw cycles. |
| Formulation | PBS and 1M Urea, pH 7.4. |
| For Use With (Application) | Blocking/Neutralizing, Control |
| Gene Alias | C6orf33Lysosomal membrane protein in brain 1, LMPB1FLJ32521, lysosomal membrane protein in brain-1, membrane progestin receptor beta, mPR beta, MPRBFLJ46206, Progestin and adipoQ receptor family member 8, progestin and adipoQ receptor family member VIIIch |
| Gene Symbol | PAQR8 |
| Label Type | Unlabeled |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
