Learn More
CPC Scientific H-Phe-Ser-Leu-Leu-Arg-Tyr-NH2 (trifluoroacetate salt) 25MG
SDP
Supplier:  CPC Scientific PARP004B
Encompass
This peptide inhibited activation of PAR-2 by trypsin in PAR-2 receptor expressing KNRK cells. Half-maximal inhibition of calcium signaling was observed at about 50 ?M. In contrast, the activation of PAR-2 by SLIGRL-NH2 (PAR-2 (1-6) amide (mouse, rat)) was not inhibited by this peptide. Selective Antagonist for PAR2 Agonist. This peptide blocks trypsin but not SLIGRL-NH2 activation of PAR2 in receptor-expressing KNRK cells.SEQUENCE: H-Phe-Ser-Leu-Leu-Arg-Tyr-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: FSLLRY-NH2MOLECULAR FORMULA: C39H60N10O8MOLECULAR WEIGHT: 796.97STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [245329-02-6]SYNONYMS: RESEARCH AREA: HematologyREFERENCES:B.Al-Ani et al., J. Pharmacol. Exp. Ther., 300, 702 (2002)S.Wilson et al., Biochem. J., 388, 967 (2005)SKU(s): PARP-004A, PARP-004B
Catalog No. 50-211-0101
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.