Learn More
CPC Scientific H-Leu-Ser-Ile-Gly-Lys-Val-NH2 (trifluoroacetate salt) 100MG
SDP
Supplier:  CPC Scientific PARP001D
Encompass
This peptide is a partially reversed human PAR-2 activating peptide serving as an inactive control for the PAR-2 agonist SLIGKV-amide.ONE-LETTER SEQUENCE: LSIGKV-NH2MOLECULAR FORMULA: C28H54N8O7MOLECULAR WEIGHT:614.79STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1348395-60-7], SYNONYMS: LSIGKVamideRESEARCH AREA: HematologyREFERENCES: H.Nishikawa et al., J. Pharmacol. Exp. Ther., 312, 324 (2005); I.A. Akers et al.,Am. J. Physiol. Lung Cell. Mol. Physiol., 278, L193 (2000); S. Miyata, et al., J. Biol. Chem., 275, 4592 (2000); S. Miike, A.S. McWilliam, and H. Kita, J. Immunol., 167, 6615 (2001)
Catalog No. 50-211-0095
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.