Learn More
CPC Scientific H-Ser-Phe-Asn-Gly-Gly-Pro-NH2 (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific PARP009A
Encompass
This peptide is a murine PAR-3-derived tethered ligand sequence which does not activate PAR-3, but rather activates PAR-1 and PAR-2, either in Jurkat T-cells or in other PAR-expressing cells. PAR3 Tethered Ligand (Murine) / Activates PAR1 and PAR2SEQUENCE: H-Ser-Phe-Asn-Gly-Gly-Pro-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: SFNGGP-NH2MOLECULAR FORMULA: C25H36N8O8MOLECULAR WEIGHT: 576.61STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [261521-21-5]SYNONYMS: Proteinase Activated Receptor 3 (1-6) amide (human), Coagulation Factor II Receptor-Like 2 (1-6) amide (human), TFRGAPamide, Thrombin Receptor-Like 2 (1-6) amide (human), TFRGAP-NH2 PAR3 Tethered Ligand (Human) / Activates PAR1 and PAR2, PAR3 (1-6) amide (mouse)RESEARCH AREA: HematologyREFERENCES:K.K.Hansen et al., Immunology, 112, 183 (2004)SKU(s): PARP-009A, PARP-009B
Catalog No. 50-211-0109
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.