Learn More
CPC Scientific H-Gly-Tyr-Pro-Gly-Lys-Phe-OH (trifluoroacetate salt) 25MG
SDP
Supplier:  CPC Scientific PARP011A
Encompass
This PAR-4 activating peptide caused secretion and aggregation of PAR-3 deficient mouse platelets.SEQUENCE: H-Gly-Tyr-Pro-Gly-Lys-Phe-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: GYPGKFMOLECULAR FORMULA: C33H45N7O8MOLECULAR WEIGHT: 667.76STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [213018-42-9]SYNONYMS: Proteinase Activated Receptor 4 (1-6) (mouse), GYPGKF, Thrombin Receptor-Like 3 (1-6) (mouse), Coagulation Factor II Receptor-Like 3 (1-6) (mouse)RESEARCH AREA: Hematology, CardiovascularREFERENCES:M.L.Kahn et al., Nature, 394, 690 (1998)SKU(s): PARP-011A
Catalog No. 50-211-0113
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.