Learn More
CPC Scientific H-Gly-Tyr-Pro-Gly-Gln-Val-OH (trifluoroacetate salt) 25MG
Supplier: CPC Scientific PARP010B
This peptide is the amino terminus of the fourth protease-activated receptor (PAR4) after the cleavage at Arg47/Gly48 was tested for its ability to stimulate COS cells expressing PAR4. The peptide readily activated both wild-type and mutant PAR4 (R47A) at 500 M, whereas thrombin and trypsin only activate the wild-type PAR4.SEQUENCE: H-Gly-Tyr-Pro-Gly-Gln-Val-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: GYPGQVMOLECULAR FORMULA: C28H41N7O9MOLECULAR WEIGHT: 619.68STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [225779-44-2]SYNONYMS: Proteinase Activated Receptor 4 (1-6) (human), GYPGQV, Thrombin Receptor-Like 3 (1-6) (human), Coagulation Factor II Receptor-Like 3 (1-6) (human)RESEARCH AREA: HematologyREFERENCES:W.-F.Xu et al., Proc. Natl. Acad. Sci. USA, 95, 6642 (1998)H.Andersen et al., Proc. Natl. Acad. Sci. USA, 96, 11189 (1999)R.Kaufmann et al., J. Cancer Res. Clin. Oncol., 126, 91 (2000)SKU(s): PARP-010A, PARP-010B
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.