Learn More
Invitrogen™ PAR2 Monoclonal Antibody (SAM11)

Description
PAR2 Monoclonal Antibody for IHC, ELISA
Specifications
Specifications
| Antigen | PAR2 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | SAM11 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.4 |
| Gene | F2rl1 |
| Gene Accession No. | P55085 |
| Gene Alias | coagulation factor II (thrombin) receptor-like 1; coagulation factor II receptor-like 1; F2R like trypsin receptor 1; F2RL1; Gpcr11; Gpr11; G-protein coupled receptor 11; PAR2; PAR-2; protease-activated receptor 2; Protease-activated receptor-2; proteinase-activated receptor 2; Proteinase-activated receptor 2, alternate cleaved 1; Proteinase-activated receptor 2, alternate cleaved 2; proteinase-activated receptor-2; Proteinase-activated receptor-2 G protein-coupled receptor 11; Proteinase-activated receptor-2, G protein-coupled receptor 11; thrombin receptor-like 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.