Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck Para 12™ Extend Hematology Control
SDP

Catalog No. 11716003 Shop All Streck Laboratories Products
Change view
Click to view available options
Level:
1 Low, 1 Normal, 1 High
6 High
6 Low
6 Low, 6 Normal, 6 High
6 Normal
Quantity:
3 x 2.5 mL
3 x 4.5 mL
6 x 2.5 mL
6 x 4.5 mL
18 x 2.5 mL
18 x 4.5 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level Quantity
11-716-003 6 Normal 6 x 4.5 mL
23-285-579 6 Low, 6 Normal, 6 High 18 x 4.5 mL
11-716-334 1 Low, 1 Normal, 1 High 3 x 2.5 mL
11-716-001 6 High 6 x 2.5 mL
23-286-109 1 Low, 1 Normal, 1 High 3 x 4.5 mL
11-716-289 6 Low, 6 Normal, 6 High 18 x 2.5 mL
11-716-000 6 Low 6 x 2.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 11-716-003 Supplier Streck Supplier No. 218995
Add to Cart
Add to Cart
This item is not returnable. View return policy

Para 12 Extend is a three-part differential hematology control assayed for Beckman Coulter™, ERMA, Horiba™ Medical, HTI™ Diagnostics, Mindray™, Nihon Kohden™ and Siemens™ 3-part hematology analyzers. It features vials with pierceable caps for autosampling and is available in low, normal and high levels.

  • Available in three clinically significant levels.
  • Plastic vials with pierceable caps for analyzer autosampling.
  • Compatible with the most popular three-part differential hematology analyzers.
  • 30-day open-vial stability, 190-day closed-vial stability.
  • Long shelf life reduces the number of shipments and associated costs.
  • Access to STATS™, our free interlaboratory quality control program for peer group data comparison.

Compliance

Complies with all applicable regulations of the U.S. Department of Transportation (DOT), and/or International Air Transportation Association (IATA).

Specifications

Format Vial
For Use With (Equipment) Abbott CELL-DYN™ 1400, 1600, 1700, and 1800, ABX micros 60, Beckman Coulter™ Ac
Type Para 12™ Extend Hematology Control
Includes Abbott CELL-DYN™ 1400, 1600, 1700, and 1800, ABX Micros 60, Beckman Coulter™ Ac•T™/Ac•T with Diff/Ac•Tdiff2, Mindray BC-3200, ERMA PCE-210, Diatron™ Abacus, Abacus Jr., Nihon Kohden Celltac and MEK-6400
Intended Use Test accuracy and precision of white cell populations
Level 6 Normal
Quantity 6 x 4.5 mL
Stability 3 Months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.