Learn More
Description
- Whole blood control resembles patient sample.
- Available in three clinically significant levels.
- Packaged in plastic squeeze dropper vials and glass vials.
- 14-day open-vial stability, 110-day closed-vial stability.
- Long shelf life reduces the number of shipments and associated costs.
- Access to STATS™, Streck’s free interlaboratory quality control program with peer group data comparison and online submission.
Specifications
Specifications
| Assay Target | WBC, RBC, Hgb, Hct and Platelet |
| Certifications/Compliance | CE |
| Content And Storage | 2°C to 10°C |
| Format | Glass Vial |
| For Use With (Equipment) | HemoCue™ B-Hemoglobin photometer and Hb201+, ITC Hgb Pro professional hemoglobin testing system |
| Type | 4 Parameter Control |
| Intended Use | For small laboratories performing a limited number of hematology tests |
| Level | 6 High |
| Quantity | 6 x 1.5 mL |
| Stability | 110 Days |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
