Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck Para 4™ Four Parameter Control
SDP

Catalog No. 23285641 Shop All Streck Laboratories Products
Change view
Click to view available options
Level:
2 Low, 2 Normal, 2 High
4 Low, 4 Normal, 4 High
6 High
6 Low
6 Low, 6 Normal, 6 High
6 Normal
Quantity:
6 x 1.5 mL
12 x 1.5 mL
18 x 1.5 mL
Type:
4 Param. Control, Glass vial
4 Param. Control, Plastic vial
4 Parameter Control
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level Quantity Type
23-285-641 2 Low, 2 Normal, 2 High 6 x 1.5 mL 4 Parameter Control
11-716-282 6 Normal 6 x 1.5 mL 4 Parameter Control
23-285-547 4 Low, 4 Normal, 4 High 12 x 1.5 mL 4 Param. Control, Plastic vial
11-716-285 6 High 6 x 1.5 mL 4 Parameter Control
11-716-284 6 Low 6 x 1.5 mL 4 Parameter Control
23-285-827 2 Low, 2 Normal, 2 High 6 x 1.5 mL 4 Param. Control, Glass vial
11-716-283 6 Low, 6 Normal, 6 High 18 x 1.5 mL 4 Parameter Control
11-716-17 4 Low, 4 Normal, 4 High 12 x 1.5 mL 4 Param. Control, Glass vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 23-285-641 Supplier Streck Supplier No. 215110

Para 4 is a hematology control for laboratories that perform a limited number of hematology tests utilizing manual or semi-manual methods. It is available in low, normal and high levels.

  • Whole blood control resembles patient sample.
  • Available in three clinically significant levels.
  • Packaged in plastic squeeze dropper vials and glass vials.
  • 14-day open-vial stability, 110-day closed-vial stability.
  • Long shelf life reduces the number of shipments and associated costs.
  • Access to STATS™, Streck’s free interlaboratory quality control program with peer group data comparison and online submission.

Specifications

Assay Target WBC, RBC, Hgb, Hct and Platelet
Certifications/Compliance CE
Content And Storage 2°C to 10°C
Format Plastic Squeeze Dropper Vial
For Use With (Equipment) HemoCue™ B-Hemoglobin photometer and Hb201+, ITC Hgb Pro professional hemoglobin testing system
Type 4 Parameter Control
Intended Use For small laboratories performing a limited number of hematology tests
Level 2 Low, 2 Normal, 2 High
Quantity 6 x 1.5 mL
Stability 110 Days
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.