Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paraffin, liquid, technical

Catalog No. AC444180250
Change view
Click to view available options
Quantity:
25 L
Packaging:
Plastic Can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC444180250 25 L Plastic Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC444180250 Supplier Thermo Scientific Chemicals Supplier No. 444180250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 8012-95-1 | MFCD00131611

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8012-95-1
Molecular Formula MFCD00131611
Molecular Weight (g/mol) 0.00
MDL Number MFCD00131611
InChI Key FFNDMZIBVDSQFI-UHFFFAOYSA-N
Synonym delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride
PubChem CID 68245
ChEBI CHEBI:38701
IUPAC Name 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
SMILES *

Specifications

Melting Point -24.0°C
Density 0.8800g/mL
Boiling Point 300.0°C
Flash Point 218°C
Packaging Plastic Can
Quantity 25 L
Specific Gravity 0.88
Solubility Information Solubility in water: insoluble
Viscosity 65-75 mm2/s (40°C)
Grade Technical
Chemical Name or Material Paraffin, liquid
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 232-384-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.