Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Paraffin Type 1, 3, 6, 9
GSA_VA

Catalog No. 22050126 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Paraffin Type 1
Paraffin Type 3
Paraffin Type 6
Paraffin Type 9
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-050-126 Paraffin Type 6
22-050-124 Paraffin Type 1
22-050-125 Paraffin Type 3
22-050-127 Paraffin Type 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-050-126 Supplier Epredia™ Supplier No. 8336
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Find the perfect fit with four different blends of highly refined paraffin and polymers, offering a choice of infiltration times and cutting characteristics.

  • All paraffins melt between 55° and 57°C and are triple filtered to 0.5μm
  • Flake form has a greater surface-to-weight ratio than pellets, resulting in a faster melting time
  • Type 1 contains no polymer and offers rapid infiltration
  • Type 3 contains minimal polymer and offers fast infiltration
  • Type 6 is harder than Type 3 and allows for thinner sections
  • Type 9 has the highest polymer content available and yields extremely thin sections

Ordering Information:
Packaged in convenient, easy-to-open, resealable bags

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Melting Point 54° to 57°C
Product Type Paraffin Type 6
Quantity 2 lb.
For Use With (Application) Infiltration and Embedding
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.