Learn More
Market Lab 4-Inch Parafilm Roll, 4 x 125
Supplier:  Market Lab 108104
Encompass_Preferred
The 4-Inch Parafilm Roll from MarketLab is a 4 inch wide roll of Parafilm laboratory film that is 125 feet long. Parafilm is a flexible, stretchable, and moldable film made of olefin that creates an airtight and watertight seal to protect contents. This Parafilm roll can be used to seal containers, cover plates, wrap hoses, secure samples, and more in laboratory settings. It stretches to conform to any shape and bonds to itself to create a secure seal. The 4 inch width allows it to be easily torn by hand without the need for cutting. This 125 foot roll provides a large supply for sealing many containers and items in the lab.
Catalog No. NC2316598
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.