Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Paraformaldehyde, 4% in PBS + Mg + EGTA

Catalog No. AAJ62478AK
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62478AK 250 mL
AAJ62478AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62478AK Supplier Thermo Scientific Chemicals Supplier No. J62478AK
  • $132.00
    $126.65 / Each of 1

    Save $5.35

    4% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

Used as a common fixative reagent for histology. It is also used to prepare pure formaldehyde. | CAS: 30525-89-4 | CH2O | 30.026 g/mol

Paraformaldehyde, 4% in PBS + Mg + EGTA is used as a common fixative reagent for histology. It is also used to prepare pure formaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30525-89-4
Molecular Formula CH2O
Molecular Weight (g/mol) 30.026
MDL Number MFCD00133991
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Physical Form Liquid
Packaging Plastic bottle
Quantity 250 mL
Solubility Information Immiscible with water.
Chemical Name or Material Paraformaldehyde
Hazard Category H317-H318-H351
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P305+P351+P338-P308+P313-P310-P333+P313-P363-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.