Learn More
Leica Biosystems Parafree Disposable Base Molds
Supplier:  Leica Biosystems 39LC7055
Encompass
Disposable Base Molds have been designed to be compatible with any currently available cassette. The orientation of the specimen is made easier due to the transparency of the mold, while its rounded corners allow for easy block removal and paraffin ribbon continuity. Built up side walls reduce risk of paraffin seepage during embedding. Available in boxes of 400 or cases of 1600.
Catalog No. 50-255-2386
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.