Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Parainfluenza Virus Type 1 Fusion Protein Monoclonal Antibody (PAR1 (251/12A))
GREENER_CHOICE

Catalog No. PIMA516282
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516282 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516282 Supplier Invitrogen™ Supplier No. MA516282
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Parainfluenza Virus Type 1 Fusion Protein Monoclonal Antibody for ICC/IF

Parainfluenza refers to a group of viruses that lead to upper and lower respiratory infections. There are four types of parainfluenza virus, all of which can cause upper respiratory infections or lower respiratory infections (pneumonia) in adults and children. The virus can cause croup,bronchiolitis,bronchitis and certain types of pneumonia. Parainfluenza Virus (PIV) is enveloped and contains single stranded negative sense RNAwith a genome of ~15,000 nucleotides and six structural proteins. The fusion (F) protein is important in aiding the fusion of the host and viral cellular membranes, eventually forming syncytia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Parainfluenza Virus Type 1 Fusion Protein
Applications Immunocytochemistry
Classification Monoclonal
Clone PAR1 (251/12A)
Concentration 6.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.5
Gene Alias canine parainfluenza virus; hPIV-1; PIV-1
Host Species Mouse
Immunogen Parainfluenza 1 fusion protein.
Purification Method Protein A
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.