Learn More
Description
beta 2-Microglobulin (B2M) forms the light chain of Class I MHC molecules. It has been measured in a variety of body fluids, including serum/plasma, saliva, CSF, and urine. B2M freely passes through the glomerular membrane, but it is nearly completely reabsorbed and degraded in proximal tubule cells. Clinically, serum B2M is elevated in rheumatoid arthritis, systemic lupus erythematosus, viral infections, and conditions associated with decreased glomerular filtration.
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Type | Control Set for ELISA Kit |
| Gene ID (Entrez) | 567 (Human); 12010 (Mouse); 24223 (Rat) |
| Intended Use | For quantitative measurement of beta 2 microglobulin levels |
| Product Line | R&D Systems |
| Product Name | Parameter Immunoassay Control Set |
| Source | Human |
| Target Gene | beta 2-Microglobulin |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
