Learn More
Apexbio Technology LLC Parathyroid hormone (1-34) (human), 5mg. Cas: 52232-67-4 MFCD: MFCD00149013. Increases blood calcium level
SDP
Supplier:  Apexbio Technology LLC A11295
Encompass_Preferred
Parathyroid hormone (1-34) (human), (C181H291N55O51S2), a peptide with the sequence H2N-SVSEIQLMHNLGKHLNSMERVEWLRKKLQDVHNF-OH, MW= 4117.72. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4688
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.