Learn More
Apexbio Technology LLC surface antigen (208-215) [Hepatitis B Virus] 5mg
SDP
Supplier:  Apexbio Technology LLC A10915
Encompass_Preferred
Surface antigen 208-215 Hepatitis B Virus is a peptide representing a defined epitope of the hepatitis B virus surface antigen (HBsAg) which is associated with host immune recognition and antibody-mediated responses The peptide encompasses antigenic determinants recognized during HBV infection facilitating studies focused on viral pathogenesis and immune interactions Surface antigen 208-215 Hepatitis B Virus exerts its biological activity through its defined antigenic sequence serving as a target for antibody detection and immunological assays Based on these properties surface antigen 208-215 Hepatitis B Virus holds research potential in virological studies immunological assay development diagnostics and vaccine research for hepatitis B virus
Catalog No. 50-000-96694
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.