Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Parathyroid Hormone Peptide, Synthetic, Human, >95% Purity, 100 μg

Catalog No. 502710914
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-271-0914 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-271-0914 Supplier Abcam Supplier No. AB42334100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic blocking peptide with >95% purity for use in blocking applications.

  • Protein Type: Synthetic
  • Applications: BL
  • Purity: >95%
  • Form: Lyophilized
  • Species Reactivity: Human

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Parathyroid Hormone (Human)
Shipping Condition Blue Ice
Substrate Properties Peptide-based Substrate
Label or Dye Unconjugated
Content And Storage Store at -20°C.
Form Lyophilized
Format Vial
Cross Reactivity Human
Quantity 100 μg
For Use With (Application) Blocking
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.