Learn More
AdipoGen Parecoxib
SDP
Supplier:  AdipoGen CDXP0167M025
Encompass_Preferred
Chemical. CAS 198470-84-7. Formula C19H18N2O4S. MW 370.4. Synthetic. Potent and selective COX-2 inhibitor. Water-soluble amide prodrug of the cyclooxygenase II COX-2 selective, non-steroidal anti-inflammatory drug NSAID valdecoxib. Has anti-inflammatory, analgesic, and antipyretic activities. Upon administration, parecoxib is hydrolyzed by hepatic carboxyesterases to its active form, valdecoxib.
Catalog No. 50-256-0213
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.