Learn More
Invitrogen™ PARL Monoclonal Antibody (4F4)

Description
Peptide Sequence: AWRGWAQRGW GCGQAWGASV GGRSCEELTA VLTPPQLLGR RFNFFIQQKC GFRKAPRKVE PRRSDPGTSG EAYKRSALIP PVEETVFYPS PYPIRSLIK.
Specifications
Specifications
| Antigen | PARL |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 4F4 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | PARL |
| Gene Accession No. | Q9H300 |
| Gene Alias | D16Ertd607e; Mitochondrial intramembrane cleaving protease PARL; mitochondrial intramembrane-cleaving protease PARL; Parl; parl {ECO:0000250; Pbeta; P-beta; presenilin associated rhomboid like; presenilin associated, rhomboid-like; presenilin associated, rhomboid-like isoform 1 preproprotein; presenilin associated, rhomboid-like preproprotein; presenilins-associated rhomboid-like protein, mitochondrial; PRO2207; Psarl; PSARL1; PSENIP2; RHBDS1; rhomboid 7 homolog 1; UniProtKB:Q9H300} |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.