Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Paromomycin Recombinant Mouse Monoclonal Antibody (2D3)

Catalog No. MA566380
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA566380 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA566380 Supplier Invitrogen™ Supplier No. MA566380
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Recombinant Monoclonal Antibody

Paromomycin Recombinant Monoclonal Antibody for ELISA

Paromomycin is an aminoglycoside antibiotic derived from Streptomyces rimosus, exhibiting broad-spectrum activity against various infections such as amoebiasis, cryptosporidiosis, giardiasis, and numerous gram-negative and gram-positive bacteria. Its structure is characterized by a complex arrangement of sugar moieties, which contribute to its binding affinity and mechanism of action, primarily targeting the bacterial ribosome to inhibit protein synthesis. Paromomycin's gene location and modifications have been explored to enhance its efficacy and overcome resistance, with targeted chemical syntheses revealing efficient modifications within the neomycin family. These modifications aim to convert neomycin and its derivatives into more potent forms, addressing the growing concern of antibiotic resistance. Paromomycin is also investigated as a selection agent in transgenic plant studies, demonstrating its versatility beyond clinical applications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Paromomycin
Applications ELISA
Classification Recombinant Monoclonal
Clone 2D3
Concentration 5.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Host Species Mouse
Immunogen Conjugate of PAM hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.