Learn More
Enzo Life Sciences PARP-1 monoclonal antibody (C-2-10) (affinity purified) (50 µg)
SDP
Supplier:  Enzo Life Sciences BMLSA2490050
Encompass_Preferred
Clone: C-2-10. Isotype: IgG1. Immunogen: Purified calf thymus poly(ADP-ribose) polymerase (PARP). Source: Purified from ascites. Species reactivity: Human, Mouse, Rat, Monkey, Hamster, Bovine. Specificity: Recognizes an epitope in the C-terminal part of the DNA binding domain of PARP. Applications: ELISA, ICC, WB. Purity: Affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-201-1778
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.