Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP1 Rabbit, anti-Human, Unconjugated, Clone: 3N19, Proteintech
SDP

Catalog No. 502256588 Shop All Proteintech Group Inc Products
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-225-6588 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-225-6588 Supplier Proteintech Group Inc Supplier No. 801741RR100UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

The PARP1 antibody from Proteintech is a rabbit recombinant antibody to a recombinant protein of human PARP1. This antibody recognizes human, mouse, rat antigen. The PARP1 antibody has been validated for the following applications: WB,ELISA analysis.

Specifications

Antigen PARP1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3N19
Concentration 250 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 142
Target Species Human, Mouse, Rat
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.