Learn More
Invitrogen™ PARP16 Polyclonal Antibody

Description
Sequence homology: Cow: 79%; Dog: 86%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 86%; Pig: 80%; Rabbit: 86%; Rat: 86%.
Specifications
Specifications
| Antigen | PARP16 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | PARP16 |
| Gene Accession No. | Q8N5Y8 |
| Gene Alias | ADP-ribosyltransferase diphtheria toxin-like 15; ARTD15; BC055447; C15orf30; C79952; LRRGT00109; mono [ADP-ribose] polymerase PARP16; PARP16; PARP-16; pART15; poly (ADP-ribose) polymerase family, member 16; poly [ADP-ribose] polymerase 16; poly(ADP-ribose) polymerase family member 16; Protein mono-ADP-ribosyltransferase PARP16; RGD1306243 |
| Gene Symbols | PARP16 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.