Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501736019 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6019 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6019 Supplier Proteintech Group Inc Supplier No. 551491AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is specific to PARP2 (N-terminal).

This gene encodes polytransferase, but lacks an N-terminal DNA binding domain which activates the C-terminal catalytic domain of poly transferase. The basic residues within the N-terminal region of this protein may bear potential DNA-binding properties, and may be involved in the nuclear and/or nucleolar targeting of the protein. Two alternatively spliced transcript variants encoding distinct isoforms have been found.

Specifications

Antigen PARP2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PARP2
Gene Accession No. O88554, Q9UGN5
Gene Alias ADPRT 2, ADPRT2, ADPRTL2, ADPRTL3, hPARP 2, pADPRT 2, PARP 2, PARP2, poly (ADP ribose) polymerase 2, Poly [ADP ribose] polymerase 2, Poly[ADP ribose] synthase 2
Gene Symbols PARP2
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10038, 11546
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.