Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PARP9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 404400
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-440-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-440-0 Supplier Invitrogen™ Supplier No. 404400
  • $479.00
    $359.65 / Each of 1

    Save $119.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PARP9 Polyclonal Antibody for IP, Western Blot

In concert with DTX3L plays a role in PARP1-dependent DNA damage repair. PARP1-dependent PARP9/BAL1-DTX3L-mediated ubiquitination promotes the rapid and specific recruitment of 53BP1/TP53BP1, UIMC1/RAP80, and BRCA1 to DNA damage sites. Involved in inducing the expression of IFN-gamma-responsive genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PARP9
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene PARP9
Gene Accession No. Q8IXQ6
Gene Alias ADP-ribosyltransferase diphtheria toxin-like 9; ARTD9; AW214463; b aggressive lymphoma protein; B aggressive lymphoma protein homolog; Bagl; Bal; BAL1; BC003281; MGC:7868; Parp9; PARP-9; poly (ADP-ribose) polymerase 9; poly (ADP-ribose) polymerase family, member 9; poly [ADP-ribose] polymerase 9; poly(ADP-ribose) polymerase family member 9; Protein mono-ADP-ribosyltransferase PARP9; RGD1307534
Gene Symbols PARP9
Host Species Rabbit
Immunogen Synthetic peptide derived from the C-terminal region of the short isoform of human BAL protein (B-aggressive lymphoma, poly (ADP-ribose) polymerase family member 9, PARP9).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83666
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.