Learn More
Parr™ 1757 Printer for Calorimeters
SDP
Catalog No. 04731159 Shop All Parr Instrument Company Products
Change view
Click to view available options
For Use With (Equipment):
Parr 6000 Series Calorimeters
Catalog No. For Use With (Equipment)
04-731-159 Parr 6000 Series Calorimeters
1 options
Catalog No. 04-731-159 Supplier Parr™ Supplier No. 334C
Add to Cart
Add to Cart
Dedicated printer for Parr 6000-series calorimeters
For use with 6000 Series Calorimeters produced prior to October 2010.
  • Compact dot-matrix printer
  • Default printer for 6000 Series Calorimeters
  • Setup for 40-column reports
  • RS232 serial connection
  • Dedicated power supply
  • Dimensions: 5.5W × 9D × 7 in.H (13.75 × 22.9 × 17.5cm)
Order Info
Refill Paper (04-731-158) and Ink Ribbon (04-731-160) are available separately.
Specifications
Product Type Printer Paper
For Use With (Equipment) Parr 6000 Series Calorimeters