Learn More
Parr™ 6510 Water Handling System
GSA_VASDP
Catalog No. 04731176 Shop All Parr Instrument Company Products
Change view
Click to view available options
For Use With (Equipment):
Parr 6200 Calorimeter
Catalog No. For Use With (Equipment)
04-731-176 Parr 6200 Calorimeter
1 options
Catalog No. 04-731-176 Supplier Parr™ Supplier No. 6510EA
Add to Cart
Add to Cart
Request a Quote
For closed loop operation of the 6200 Calorimeter
The 6510 Water Handling System operates on the bench top, next to the calorimeter. It provides a convenient means for filling the bucket in the calorimeter with the same amount of temperature controlled water for each test run. It also serves as a source of temperature controlled water for maintaining a constant jacket temperature in the 6200 Calorimeter.
  • 5 gallon, temperature controlled reservoir
  • Built-in circulating pump
  • Attached 2000mL glass delivery pipette
  • Provides continuous filtration of all water in system
  • Built in thermoelectric cooling
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Water Handling System
For Use With (Application) Provides a convenient means for filling the bucket in the calorimeter with the same amount of temperature controlled water for each test run
For Use With (Equipment) Parr 6200 Calorimeter
Includes 5 gal. temperature controlled reservoir, built-in circulating pump, attached 2000mL glass delivery pipette, built-in thermoelectric cooler
Voltage 115/230 V