Learn More
Parr™ 6775 Digital Thermometer
SDP
Catalog No. 04731200 Shop All Parr Instrument Company Products
Change view
Click to view available options
For Use With (Equipment):
Parr 1341 Plain Jacket Calorimeter
Catalog No. For Use With (Equipment)
04-731-200 Parr 1341 Plain Jacket Calorimeter
1 options
Catalog No. 04-731-200 Supplier Parr™ Supplier No. 6775
Add to Cart
Add to Cart
Request a Quote
Precision and reliability
Displays temperature and a free running timer that can be used to aid the user in recording temperatures at regular intervals. Optimized for calorimetric temperature measurements, and especially designed to be used with the Parr 1341 Plain Jacket Calorimeter.
  • Easy to read, two-line LCD display
  • Temperature resolution: 0.001°C
  • Working temperature range: 10° to 40°C
  • Powered via two high-power lithium batteries (included)
1168E2 Thermistor Probe
  • Length: 30cm
  • 3.18mm diameter stainless-steel sheath
Specifications
Product Type Digital Thermometer
For Use With (Application) Calorimetric temperature measurements
For Use With (Equipment) Parr 1341 Plain Jacket Calorimeter
Includes Two high power lithium batteries (200 Hours)