Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parr™ Combustion Capsule for Parr Calorimeter Oxygen Combustion Bomb
SDP

Catalog No. 043855 Shop All Parr Instrument Company Products
Change view
Click to view available options
For Use With (Equipment):
Parr Calorimeter Oxygen Combustion Bomb
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
04-385-5 Parr Calorimeter Oxygen Combustion Bomb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-385-5 Supplier Parr™ Supplier No. 43AS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Parr calorimeter oxygen combustion bomb.

  • Stainless-steel
  • Dia.: approx. 1 in.
  • Depth: approx. 1cm

Specifications

Product Type Combustion Capsule
Depth (English) 0.393 in.
Depth (Metric) 1 cm
For Use With (Equipment) Parr Calorimeter Oxygen Combustion Bomb
Material Stainless Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.