Learn More
Parr™ Gelatin Capsule For Calorimeter
SDP
Catalog No. 04465 Shop All Parr Instrument Company Products
Change view
Click to view available options
For Use With (Equipment):
Parr Calorimeter
Catalog No. For Use With (Equipment)
04-465 Parr Calorimeter
1 options
Catalog No. 04-465 Supplier Parr™ Supplier No. 3601
Add to Cart
Add to Cart
Weighs and Handles Volatile Liquid Samples to be Burned in Oxygen Bomb
  • Two-Piece
  • Size 00
  • Holds 0.9mL
Specifications
Product Type Gelatin Capsule
For Use With (Application) Volatile liquid samples to be burned in an oxygen bomb can be weighed and handled conveniently in these two piece capsules
For Use With (Equipment) Parr Calorimeter