Learn More
Parr™ Ignition Wire For Calorimeter
SDP
Catalog No. 04396 Shop All Parr Instrument Company Products
Change view
Click to view available options
For Use With (Equipment):
Parr Calorimeter
Catalog No. For Use With (Equipment)
04-396 Parr Calorimeter
1 options
Catalog No. 04-396 Supplier Parr™ Supplier No. 45C10
Add to Cart
Add to Cart
Ease in cutting fuse to length
  • Nickel alloy fuse wire
  • 10cm card
  • 250 pieces
Specifications
Product Type Ignition Wire
For Use With (Application) Ease in cutting a fuse to length
For Use With (Equipment) Parr Calorimeter
Includes 10cm card
Material Nickel-Chromium Alloy