Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parter Medical Products™ Specialty Space Saver Petri Dishes

Catalog No. 08757010 Shop All Parter Medical Products Products
Change view
Click to view available options
Diameter (Metric):
100 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
08-757-010 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-757-010 Supplier Parter Medical Products™ Supplier No. 3511N

Engineered for optimum flatness to provide uniform agar thickness

  • Made from pure virgin polystyrene
  • Provides optical clarity, permitting unimpaired observation of dish content for micro photography
  • Designed with nesting lugs for optimum culture growth and media shelf life
  • Undivided bottom

Specifications

Material Polystyrene
Shape Undivided Bottom
Sterility Sterile
For Use With (Application) For optimum flatness to provide uniform agar thickness
Height (Metric) 10 mm
Diameter (Metric) 100 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.