Learn More
Apexbio Technology LLC Bioymifi 1420071-30-2 10mg
SDP
Supplier:  Apexbio Technology LLC B878510
Encompass_Preferred
Bioymifi (CAS number 1420071-30-2) is a synthetic small molecule functioning as a tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) mimetic Its biological mechanism involves selective interaction with death receptor 5 (DR5) initiating apoptotic signaling cascades potentially via caspase activation Ongoing studies explore bioymifi s suitability as an experimental anti-cancer compound targeting apoptosis-resistant tumor cell lines and elucidating DR5-mediated signaling pathways
Catalog No. 50-000-63289
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.